Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Morphine Mouse Monoclonal Antibody [clone: BDI918]

Supplier: Genetex
GTX40331
89336-930
Anti-Morphine Mouse Monoclonal Antibody [clone: BDI918]
Type: Primary
Antigen: Morphine
Clonality: Monoclonal
Clone: BDI918
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty