Anti-TRP1 Mouse Monoclonal Antibody [clone: 402]

Supplier: GeneTex
GTX40314
89336-646EA 804.8 USD
89336-646
Anti-TRP1 Mouse Monoclonal Antibody [clone: 402]
Antibodies
Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: 402
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR