Anti-IgM µ Mouse Monoclonal Antibody [clone: 47.2.E9]

Supplier: GeneTex
GTX40297
89299-554EA 655.29 USD
89299-554
Anti-IgM µ Mouse Monoclonal Antibody [clone: 47.2.E9]
Antibodies
Mouse monoclonal antibody [47.2.E9] to IgM

Recommended Starting Dilutions:ELISA: 1/5000 - 1/10000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IgM µ
Clonality: Monoclonal
Clone: 47.2.E9
Conjugation:
Epitope: µ-chain
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR