Anti-IgM µ Rat Monoclonal Antibody [clone: HM-7]

Supplier: GeneTex
GTX40291
89275-384EA 822.13 USD
89275-384
Anti-IgM µ Rat Monoclonal Antibody [clone: HM-7]
Antibodies
Rat Monoclonal antibody [HM-7] to Human IgM Mu chain

Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgM µ
Clonality: Monoclonal
Clone: HM-7
Conjugation:
Epitope: µ-chain
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR