Anti-Bacterial Luciferase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: GeneTex
GTX40289
89336-708EA 762.27 USD
89336-708
Anti-Bacterial Luciferase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: Bacterial Luciferase
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR