Anti-PAI1 Mouse Monoclonal Antibody [clone: 3E529]

Supplier: GeneTex
GTX40286
89286-442EA 640.4 USD
89286-442
Anti-PAI1 Mouse Monoclonal Antibody [clone: 3E529]
Antibodies
Mouse monoclonal antibody [3E529] to PAI-1

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PAI1
Clonality: Monoclonal
Clone: 3E529
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR