Anti-Immunodeficiency Virus 1 P24 Mouse Monoclonal Antibody [clone: 28]

Supplier: GeneTex
GTX40278
89276-110EA 885.89 USD
89276-110
Anti-Immunodeficiency Virus 1 P24 Mouse Monoclonal Antibody [clone: 28]
Antibodies
Type: Primary
Antigen: Immunodeficiency Virus 1 P24
Clonality: Monoclonal
Clone: 28
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR