Anti-IgA Mouse Monoclonal Antibody [clone: MARA-2]

Supplier: GeneTex
GTX40277
89276-338EA 774.8 USD
89276-338
Anti-IgA Mouse Monoclonal Antibody [clone: MARA-2]
Antibodies
Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone: MARA-2
Conjugation:
Epitope: Heavy chain
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR