Anti-Infectious Rhinotracheitis Mouse Monoclonal Antibody [clone: BEL 051]

GTX40273
89299-546EA 360.48 USD
89299-546
Anti-Infectious Rhinotracheitis Mouse Monoclonal Antibody [clone: BEL 051]
Type: Primary
Antigen: Infectious Rhinotracheitis
Clonality: Monoclonal
Clone: BEL 051
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR