Anti-Influenza A Mouse Monoclonal Antibody [clone: A1 and A3]

Supplier: GeneTex
GTX40270
89265-022EA 1216.29 USD
89265-022
Anti-Influenza A Mouse Monoclonal Antibody [clone: A1 and A3]
Antibodies
Type: Primary
Antigen: Influenza A
Clonality: Monoclonal
Clone: A1 and A3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR