Anti-Influenza A H1 Antigen Mouse Monoclonal Antibody [clone: 58AB7-19-18]

Supplier: GeneTex
GTX40268
89262-396EA 1070.81 USD
89262-396
Anti-Influenza A H1 Antigen Mouse Monoclonal Antibody [clone: 58AB7-19-18]
Antibodies
Type: Primary
Antigen: Influenza A H1 Antigen
Clonality: Monoclonal
Clone: 58AB7-19-18
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR