Anti-Influenza A H1 Antigen Mouse Monoclonal Antibody [clone: 58CE8-1-5]

GTX40266
89262-394EA 442.44 USD
89262-394
Anti-Influenza A H1 Antigen Mouse Monoclonal Antibody [clone: 58CE8-1-5]
Type: Primary
Antigen: Influenza A H1 Antigen
Clonality: Monoclonal
Clone: 58CE8-1-5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR