Anti-Influenza A H1N1 Mouse Monoclonal Antibody [clone: 9B3.2]

Supplier: GeneTex
GTX40264
89265-020EA 1044.92 USD
89265-020
Anti-Influenza A H1N1 Mouse Monoclonal Antibody [clone: 9B3.2]
Antibodies
Type: Primary
Antigen: Influenza A H1N1
Clonality: Monoclonal
Clone: 9B3.2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR