Anti-Influenza A H5 Antigen Mouse Monoclonal Antibody [clone: 11A9]

GTX40256
89279-348EA 441.78 USD
89279-348
Anti-Influenza A H5 Antigen Mouse Monoclonal Antibody [clone: 11A9]
Type: Primary
Antigen: Influenza A H5 Antigen
Clonality: Monoclonal
Clone: 11A9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR