Anti-Influenza A Nucleoprotein Mouse Monoclonal Antibody [clone: F8]

Supplier: GeneTex
GTX40249
89265-870EA 656.38 USD
89265-870
Anti-Influenza A Nucleoprotein Mouse Monoclonal Antibody [clone: F8]
Antibodies
Type: Primary
Antigen: Influenza A Nucleoprotein
Clonality: Monoclonal
Clone: F8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR