Anti-RAD51D Mouse Monoclonal Antibody [clone: Rad51D 5B3/6]

Supplier: GeneTex
GTX40243
89328-624EA 482.52 USD
89328-624
Anti-RAD51D Mouse Monoclonal Antibody [clone: Rad51D 5B3/6]
Antibodies
Mouse monoclonal [Rad51D 5B3/6] to Rad51D

WB: Use at an assay dependent dilution. Predicted molecular weight: 35 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RAD51D
Clonality: Monoclonal
Clone: Rad51D 5B3/6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR