Anti-RAD51B Mouse Monoclonal Antibody [clone: Rad51B 1H3/13]

Supplier: GeneTex
GTX40242
89328-622EA 630.11 USD
89328-622
Anti-RAD51B Mouse Monoclonal Antibody [clone: Rad51B 1H3/13]
Antibodies
Mouse monoclonal [Rad51B 1H3/13] to Rad51B

WB: Use at an assay dependent dilution. Predicted molecular weight: 38 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RAD51B
Clonality: Monoclonal
Clone: Rad51B 1H3/13
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR