Anti-Mycobacterium tuberculosis CFP10 (10kDa culture filtrate protein) Mouse Monoclonal Antibody [clone: B386M]

Supplier: GeneTex
GTX40241
89295-158EA 498.3 USD
89295-158
Anti-Mycobacterium tuberculosis CFP10 (10kDa culture filtrate protein) Mouse Monoclonal Antibody [clone: B386M]
Antibodies
Type: Primary
Antigen: Mycobacterium tuberculosis CFP10 (10kDa culture filtrate protein)
Clonality: Monoclonal
Clone: B386M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR