Anti-Mycobacterium tuberculosis ESAT6 (6kDa early secretory antigen of T cells) Mouse Monoclonal Antibody [clone: B392M]

Supplier: GeneTex
GTX40232
89295-152EA 501.76 USD
89295-152
Anti-Mycobacterium tuberculosis ESAT6 (6kDa early secretory antigen of T cells) Mouse Monoclonal Antibody [clone: B392M]
Antibodies
Type: Primary
Antigen: Mycobacterium tuberculosis ESAT6 (6kDa early secretory antigen of T cells)
Clonality: Monoclonal
Clone: B392M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR