Anti-Influenza B Nucleoprotein Mouse Monoclonal Antibody [clone: B114]

Supplier: GeneTex
GTX40229
89261-128EA 986.79 USD
89261-128
Anti-Influenza B Nucleoprotein Mouse Monoclonal Antibody [clone: B114]
Antibodies
Type: Primary
Antigen: Influenza B Nucleoprotein
Clonality: Monoclonal
Clone: B114
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR