Anti-Hairy Mouse Monoclonal Antibody [clone: 39470]

Supplier: GeneTex
GTX40165
89328-604EA 482.52 USD
89328-604
Anti-Hairy Mouse Monoclonal Antibody [clone: 39470]
Antibodies
Type: Primary
Antigen: Hairy
Clonality: Monoclonal
Clone: 39470
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR