Anti-Rubella virus core antigen Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

GTX40114
89291-632EA 399.48 USD
89291-632
Anti-Rubella virus core antigen Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: Rubella virus core antigen
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR