Anti-Mycobacterium tuberculosis Mouse Monoclonal Antibody [clone: BDI578]

Supplier: GeneTex
GTX40045
89262-386EA 643.88 USD
89262-386
Anti-Mycobacterium tuberculosis Mouse Monoclonal Antibody [clone: BDI578]
Antibodies
Type: Primary
Antigen: Mycobacterium tuberculosis
Clonality: Monoclonal
Clone: BDI578
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR