Anti-Chlamydia trachomatis Mouse Monoclonal Antibody [clone: 130]

Supplier: GeneTex
GTX40037
89280-204EA 602.91 USD
89280-204
Anti-Chlamydia trachomatis Mouse Monoclonal Antibody [clone: 130]
Antibodies
Type: Primary
Antigen: Chlamydia trachomatis
Clonality: Monoclonal
Clone: 130
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR