Anti-IGF1 Mouse Monoclonal Antibody [clone: M23]

Supplier: GeneTex
GTX39997
89276-336EA 888.97 USD
89276-336
Anti-IGF1 Mouse Monoclonal Antibody [clone: M23]
Antibodies
Mouse monoclonal antibody [M23] to IGF1

Type: Primary
Antigen: IGF1
Clonality: Monoclonal
Clone: M23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR