Anti-ITGA9 Mouse Monoclonal Antibody [clone: Y9A2]

Supplier: GeneTex
GTX39988
89262-384EA 612.2 USD
89262-384
Anti-ITGA9 Mouse Monoclonal Antibody [clone: Y9A2]
Antibodies
Mouse monoclonal antibody [Y9A2] to Integrin alpha 9 + beta 1

Type: Primary
Antigen: ITGA9
Clonality: Monoclonal
Clone: Y9A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR