Anti-IFNB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX39984
89262-376EA 660.25 USD
89262-376
Anti-IFNB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IFN beta

Type: Primary
Antigen: IFNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR