Anti-IFNG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX39983
89262-374EA 660.25 USD
89262-374
Anti-IFNG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IFN gamma

Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR