Anti-IFNG Mouse Monoclonal Antibody [clone: G-23]

GTX39973
89280-506EA 455.89 USD
89280-506
Anti-IFNG Mouse Monoclonal Antibody [clone: G-23]
Mouse monoclonal antibody [G-23] to IFN gamma

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: G-23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR