Anti-IL10 Mouse Monoclonal Antibody [clone: CC320]

Supplier: GeneTex
GTX39963
89274-884EA 1027.39 USD
89274-884
Anti-IL10 Mouse Monoclonal Antibody [clone: CC320]
Antibodies
Mouse monoclonal antibody [CC320] to IL10

Recommended Starting Dilutions:ELISA: 5 ug/ml - 10 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: CC320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Goat, Horse, Sheep, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR