Anti-IL10 Rat Monoclonal Antibody [clone: JES3-12G8]

Supplier: GeneTex
GTX39962
89275-370EA 756.79 USD
89275-370
Anti-IL10 Rat Monoclonal Antibody [clone: JES3-12G8]
Antibodies
Rat monoclonal antibody [JES3-12G8] to IL10

Recommended Starting Dilutions:Western Blotting: 1/100 - 1/250Immunohistology - Paraffin: Max Dilution 1/100Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: JES3-12G8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR