Anti-IL12B Mouse Monoclonal Antibody [clone: CC326]

Supplier: GeneTex
GTX39959
89333-488EA 653.98 USD
89333-488
Anti-IL12B Mouse Monoclonal Antibody [clone: CC326]
Antibodies
Mouse monoclonal antibody [CC326] to IL12

Recommended Starting Dilutions:ELISA: 1/100 - 1/1000Functional Assays: Max Dilution 1/100Flow Cytometry: Max Dilution 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL12B
Clonality: Monoclonal
Clone: CC326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Sheep, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR