Anti-Mycobacterium tuberculosis 38kDa Mouse Monoclonal Antibody [clone: HTM82]

Supplier: GeneTex
GTX39955
89295-124EA 516.82 USD
89295-124
Anti-Mycobacterium tuberculosis 38kDa Mouse Monoclonal Antibody [clone: HTM82]
Antibodies
Type: Primary
Antigen: Mycobacterium tuberculosis 38kDa
Clonality: Monoclonal
Clone: HTM82
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR