Anti-IL2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX39952
89262-360EA 826.2 USD
89262-360
Anti-IL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IL2

Recommended Starting Dilutions:ELISA: 0.5 - 2.0 ug/mlFunctional Assays: 0.12 - 0.16 ug/mlWestern Blotting: 0.1 - 0.2 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR