Anti-IL4 Mouse Monoclonal Antibody [clone: CC303]

Supplier: GeneTex
GTX39941
89333-486EA 653.98 USD
89333-486
Anti-IL4 Mouse Monoclonal Antibody [clone: CC303]
Antibodies
Mouse monoclonal antibody [CC303] to IL4

Recommended Starting Dilutions:Flow Cytometry: 1/50 - 1/100Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: CC303
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Pig, Dolphin, Goat, Horse, Mink, Sheep, Bovine, Mustelid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR