Anti-Interleukin-6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX39935
89280-852EA 800.02 USD
89280-852
Anti-Interleukin-6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Interleukin-6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR