Anti-MPO Mouse Monoclonal Antibody [clone: 17G2]

Supplier: GeneTex
GTX39924
89295-120EA 610.67 USD
89295-120
Anti-MPO Mouse Monoclonal Antibody [clone: 17G2]
Antibodies
Mouse Monoclonal antibody [17G2] to Myeloperoxidase

Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: 17G2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR