Anti-Ig kappa Goat Polyclonal Antibody

Supplier: GeneTex
GTX39909
89282-750EA 504.35 USD
89282-750
Anti-Ig kappa Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Human kappa Light chain

Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: Igkappa
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR