Anti-Ig kappa Goat Polyclonal Antibody

Supplier: GeneTex
GTX39908
89265-858EA 551.12 USD
89265-858
Anti-Ig kappa Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Igkappa
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR