Anti-FGF7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX39903
89276-334EA 843.63 USD
89276-334
Anti-FGF7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to KGF

Recommended Starting Dilutions:ELISA: Max Dilution 0.5 ug/mlFunctional Assays: 0.56 - 0.85 ug/mlWestern Blotting: 0.1 - 0.2 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: FGF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR