Anti-Newcastle Disease Virus (NDV) Mouse Monoclonal Antibody [clone: B535M]

Supplier: GeneTex
GTX39896
89288-208EA 540.61 USD
89288-208
Anti-Newcastle Disease Virus (NDV) Mouse Monoclonal Antibody [clone: B535M]
Antibodies
Type: Primary
Antigen: Newcastle Disease Virus (NDV)
Clonality: Monoclonal
Clone: B535M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR