Anti-KIR Mouse Monoclonal Antibody [clone: NKVFS1]

Supplier: GeneTex
GTX39888
89333-702EA 255.69 USD
89333-702
Anti-KIR Mouse Monoclonal Antibody [clone: NKVFS1]
Antibodies
Mouse monoclonal antibody [NKVFS1] to KIR family

Recommended Starting Dilutions:Flow Cytometry: 1/50 - 1/100Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: KIR
Clonality: Monoclonal
Clone: NKVFS1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR