Anti-Norovirus capsid protein Mouse Monoclonal Antibody [clone: B114M]

Supplier: GeneTex
GTX39875
89275-360EA 577.03 USD
89275-360
Anti-Norovirus capsid protein Mouse Monoclonal Antibody [clone: B114M]
Antibodies
Type: Primary
Antigen: Norovirus capsid protein
Clonality: Monoclonal
Clone: B114M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR