Anti-IgL Goat Polyclonal Antibody

GTX39864
89299-082EA 376.99 USD
89299-082
Anti-IgL Goat Polyclonal Antibody
Goat polyclonal antibody to Lambda light chain

Type: Primary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR