Anti-IgL Goat Polyclonal Antibody

Supplier: GeneTex
GTX39863
89284-448EA 421.14 USD
89284-448
Anti-IgL Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Human lambda Light chain

Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR