Anti-Legionella Pneumophila Serogroups 1/9 Mouse Monoclonal Antibody [clone: LP8]

GTX39842
89280-500EA 466.36 USD
89280-500
Anti-Legionella Pneumophila Serogroups 1/9 Mouse Monoclonal Antibody [clone: LP8]
Type: Primary
Antigen: Legionella Pneumophila Serogroups 1/9
Clonality: Monoclonal
Clone: LP8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR