Anti-Leishmania Major Surface Protease gp63 Mouse Monoclonal Antibody [clone: 96-126]

Supplier: GeneTex
GTX39837
89333-606EA 630.12 USD
89333-606
Anti-Leishmania Major Surface Protease gp63 Mouse Monoclonal Antibody [clone: 96-126]
Antibodies
Type: Primary
Antigen: Leishmania Major Surface Protease gp63
Clonality: Monoclonal
Clone: 96-126
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR