Anti-PENK Mouse Monoclonal Antibody [clone: NOC.1]

Supplier: GeneTex
GTX39835
89280-202EA 463.35 USD
89280-202
Anti-PENK Mouse Monoclonal Antibody [clone: NOC.1]
Mouse monoclonal antibody [NOC.1] to Leu-Enkephalin

Recommended Starting Dilutions:Immunohistology - Frozen: 1/100 - 1/400Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: PENK
Clonality: Monoclonal
Clone: NOC.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR