Anti-Salmonella species (A B C D E F & G Groups) Mouse Monoclonal Antibody [clone: B343M]

Supplier: GeneTex
GTX39182
89279-260EA 636.85 USD
89279-260
Anti-Salmonella species (A B C D E F & G Groups) Mouse Monoclonal Antibody [clone: B343M]
Antibodies
Type: Primary
Antigen: Salmonella species (A B C D E F & G Groups)
Clonality: Monoclonal
Clone: B343M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR