Anti-NFASC Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37602
89328-532EA 586.76 USD
89328-532
Anti-NFASC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Neurofascin 155

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: NFASC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken, Dog, Rat, Horse, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR